missing translation for 'onlineSavingsMsg'
Learn More

Racepinephrine Hydrochloride, USP, 97-102%, Spectrum™ Chemical

Catalog No. p-7158128
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
100 g
500 g
1 kg
Packaging:
Amber Glass Bottle
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18606029 1 g Amber Glass Bottle
18606030 5 g Amber Glass Bottle
18606034 25 g Amber Glass Bottle
18606031 100 g Amber Glass Bottle
18606032 500 g Amber Glass Bottle
18606033 1 kg Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. 18606029 Supplier Spectrum Chemical Mfg Cor Supplier No. E14181GM
Only null left

E1418, 329-63-5, C9H13NO3·HCl

Spectrum™ Chemical Racepinephrine Hydrochloride, USP is a bronchodilator used to relieve occasional symptoms of asthma, such as wheezing, chest tightness, and feeling short of breath. All Spectrum Chemical USP products, including all USP hydrochloride products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 329-63-5
Molecular Formula C9H14ClNO3
Molecular Weight (g/mol) 219.67
InChI Key ATADHKWKHYVBTJ-UHFFFAOYNA-N
IUPAC Name hydrogen 4-[1-hydroxy-2-(methylamino)ethyl]benzene-1,2-diol chloride
SMILES [H+].[Cl-].CNCC(O)C1=CC=C(O)C(O)=C1

Specifications

CAS Max % 1
Ignition Residue 0.005
Packaging Amber Glass Bottle
Quantity 1 g
Specific Rotation -1° to +1°
Percent Purity 97 to 102%
Grade USP
Physical Form Crystalline Powder or Lumps