Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Rabeprazole Sulfide 98.0+%, TCI America™
Specifications
Specifications
| Melting Point | 116°C |
| Color | White-Yellow |
| Quantity | 200 mg |
| Formula Weight | 343.45 |
| Percent Purity | ≥98.0% (HPLC,T) |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | Rabeprazole Sulfide |
Safety and Handling
| TSCA | No |