missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IgG Rabbit anti-Sheep, FITC, Polyclonal, Secondary Antibody, MilliporeSigma™
Description
Rabbit anti-Sheep IgG Antibody, FITC conjugate is an antibody against Sheep IgG for use in immunofluoresence.
Spécifications
Spécifications
| Antigène | Rabbit anti-Sheep IgG |
| Applications | Immunofluorescence |
| Classification | Polyclonal |
| Conjugué | FITC |
| Formule | Lyophilized from 0.02M Phosphate buffer, 0.25M NaCl with 15mg/mL BSA and 0.1% sodium azide. RECONSTITUTION: Reconstitute to 1mg/mL with sterile distilled water. |
| Espèces hôtes | Rabbit |
| Immunogène | Purified Sheep IgG. |
| Quantité | 1.5 mg |
| Primaire ou secondaire | Secondary |
| Espèces cibles | Sheep |
| Afficher plus de résultats |
For Research Use Only