missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IgG Rabbit anti-Mouse, HRP, Polyclonal, Secondary Antibody, MilliporeSigma™
Description
This Rabbit anti-Mouse IgG Antibody, HRP conjugate is validated for use in ELISA, western blotting for the detection of Mouse IgG.
Spécifications
Spécifications
| Antigène | Rabbit anti-Mouse IgG |
| Applications | ELISA, Western Blot |
| Classification | Polyclonal |
| Conjugué | HRP |
| Espèces hôtes | Rabbit |
| Quantité | 1.5 mL |
| Primaire ou secondaire | Secondary |
| Espèces cibles | Mouse |
For Research Use Only