Learn More
Rabbit anti-Mouse IgG (H+L), Superclonal™ Recombinant Secondary Antibody, HRP, Invitrogen™
Description
The sensitivity and specificity of each lot is confirmed using ELISA. Minimal cross-reactivity with goat, rat, human, bovine, guinea pig, and donkey IgG is observed. Recombinant rabbit polyclonal antibodies are unique offerings from Thermo Fisher Scientific. They are comprised of a selection of multiple different recombinant monoclonal antibodies, providing the best of both worlds – the sensitivity of polyclonal antibodies with the specificity of monoclonal antibodies - all delivered with the consistency only found in a recombinant antibody. While functionally the same as a polyclonal antibody – recognizing multiple epitope sites on the target and producing higher detection sensitivity for low abundance targets – a recombinant rabbit polyclonal antibody has a known mixture of light and heavy chains. The exact population can be produced in every lot, circumventing the biological variability typically associated with polyclonal antibody production.
Spécifications
Spécifications
| Antigène | Mouse IgG (H+L) |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugué | HRP |
| Espèces hôtes | Rabbit |
| Immunogène | Recombinant full-length protein |
| Méthode de purification | Gravity column chromatography |
| Statut réglementaire | RUO |
| Primaire ou secondaire | Secondary |
| Espèces cibles | Mouse |
| Afficher plus de résultats |