missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Rabbit anti-Mouse IgG Fc Superclonal™ Secondary Antibody

Numéro de catalogue. SA510390
Modifier l'affichage
Click to view available options
Quantité:
250 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantité
SA510390 250 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Numéro de catalogue. SA510390 Fournisseur Invitrogen™ Code fournisseur SA510390

Pour de l'aide SVP utiliser la function de clavardage; nous envoyer un couriel à help@thermofisher.com ou appelez le service à la clientèle au 1-800-234-7437.

Rabbit Recombinant Superclonal Secondary Antibody

Rabbit anti-Mouse IgG Fc Superclonal™ Recombinant Secondary Antibody is specific to Fc fragment of Mouse IgG. It can detect all subtypes of Mouse IgG (Mouse IgG1,2a,2b and 3). It has <15% cross reactivity to Human IgG. It showed minimum to no cross-reactivity to various species immunoglobulins tested. This recombinant antibody has been engineered with point mutations that eliminate binding to Fc receptors, enabling reduction in background and superior signal to noise. Superclonal™ recombinant secondary antibodies are produced using a multifaceted clonal selection process that involves several phenotypic screens to ensure enhanced specificity, animal origin free formulation and lot-to-lot consistency. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest.

Anti-Mouse secondary antibodies are affinity-purified antibodies with well-characterized specificity for mouse immunoglobulins and are useful in the detection, sorting or purification of its specified target. Secondary antibodies offer increased versatility enabling users to use many detection systems (e.g. HRP, AP, fluorescence). They can also provide greater sensitivity through signal amplification as multiple secondary antibodies can bind to a single primary antibody. Most commonly, secondary antibodies are generated by immunizing the host animal with a pooled population of immunoglobulins from the target species and can be further purified and modified (i.e. immunoaffinity chromatography, antibody fragmentation, label conjugation, etc.) to generate highly specific reagents.

Spécifications

Antigène Mouse IgG Fc
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Recombinant Superclonal
Concentration 1 mg/mL
Conjugué Unconjugated
Formule PBS with 0.09% sodium azide; pH 7.2-7.4
Espèces hôtes Rabbit
Immunogène Purified Mouse IgG.
Méthode de purification Affinity chromatography
Quantité 250 μL
Statut réglementaire RUO
Primaire ou secondaire Secondary
Espèces cibles Mouse
Contenu et stockage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Type de produit Secondary Antibody
Forme Liquid
Isotype IgG
Afficher plus de résultats Afficher moins de résultats