missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Rabbit anti-Mouse IgG Fc Secondary Antibody

Numéro de catalogue. PI31194
Modifier l'affichage
Click to view available options
Quantité:
2 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantité
PI31194 2 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Numéro de catalogue. PI31194 Fournisseur Invitrogen™ Code fournisseur 31194
  • ONLINE EXCLUSIVE PRICE
    $558.60
    $323.25 / chaque

    Économisez $235.35

    42.0 % de rabais Les écarts et divulgations suivants peuvent entraîner des prix inférieurs aux rabais indiqués, sans toutefois entraîner de réductions de prix supplémentaires : Le prix affiché reflète une réduction promotionnelle temporaire. Aucun achat additionnel ni engagement commercial n’est requis pour bénéficier du prix réduit. Le prix peut être modifié en tout temps. Le prix promotionnel est valide uniquement pour les commandes en ligne. Votre tarification contractuelle peut différer. D’autres exclusions peuvent s’appliquer.

Pour de l'aide SVP utiliser la function de clavardage; nous envoyer un couriel à help@thermofisher.com ou appelez le service à la clientèle au 1-800-234-7437.

Rabbit Polyclonal Secondary Antibody

Concentration may vary slightly from lot-to-lot, see lot-specific datasheet for exact concentration. Product # 31194 has been successfully used in Western blot, IF, ICC, IHC, IP and FACS applications. Product # 31194 reacts with the heavy chains of mouse IgG but not with the light chains of most mouse immunoglobulins. This antibody does not react against mouse IgM or non-immunoglobulin serum proteins. However, this antibody may cross-react with immunoglobulins from other species. Store product at 4°C until opened. After opening, dilute only enough antibody for a single day's use. Store remainder at 4°C under sterile conditions. Country of Origin: USA.

Anti-Mouse secondary antibodies are affinity-purified antibodies with well-characterized specificity for mouse immunoglobulins and are useful in the detection, sorting or purification of its specified target. Secondary antibodies offer increased versatility enabling users to use many detection systems (e.g. HRP, AP, fluorescence). They can also provide greater sensitivity through signal amplification as multiple secondary antibodies can bind to a single primary antibody. Most commonly, secondary antibodies are generated by immunizing the host animal with a pooled population of immunoglobulins from the target species and can be further purified and modified (i.e. immunoaffinity chromatography, antibody fragmentation, label conjugation, etc.) to generate highly specific reagents.

Spécifications

Antigène Mouse IgG Fc
Applications Flow Cytometry, Immunohistochemistry, Immunoprecipitation, Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 2.3 mg/mL
Conjugué Unconjugated
Formule PBS with no preservative; pH 7.6
Espèces hôtes Rabbit
Méthode de purification Antigen affinity chromatography
Quantité 2 mg
Statut réglementaire RUO
Primaire ou secondaire Secondary
Espèces cibles Mouse
Contenu et stockage 4°C
Type de produit Secondary Antibody
Forme Liquid
Isotype IgG
Afficher plus de résultats Afficher moins de résultats