missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IgG Rabbit anti-Mouse, Cy3, Polyclonal, Secondary Antibody, MilliporeSigma™
Description
Detect Mouse IgG using this Rabbit anti-Mouse IgG Antibody, Cy3 conjugate validated for use in immunofluoresence.
Spécifications
Spécifications
| Antigène | Rabbit anti-Mouse IgG |
| Applications | Immunofluorescence |
| Classification | Polyclonal |
| Concentration | 1.4mg/mL |
| Conjugué | Cyanine3 |
| Espèces hôtes | Rabbit |
| Immunogène | Highly purified Mouse IgG isolated from pooled normal Mouse Serum. |
| Quantité | 1.5 mg |
| Primaire ou secondaire | Secondary |
| Espèces cibles | Mouse |
| Afficher plus de résultats |
For Research Use Only