missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IgG Rabbit anti-Human, Polyclonal, Secondary Antibody, MilliporeSigma™
Description
This Rabbit anti-Human IgG Antibody is validated for use in ELISA, immunoprecipitation, western blotting for the detection of Human IgG.
Spécifications
Spécifications
| Antigène | Rabbit anti-Human IgG Antibody |
| Applications | ELISA, Immunoprecipitation, Western Blot |
| Classification | Polyclonal |
| Conjugué | Unconjugated |
| Espèces hôtes | Rabbit |
| Quantité | 2 mg |
| Primaire ou secondaire | Secondary |
| Espèces cibles | Human |
| Forme | Liquid |
For Research Use Only