missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Rabbit anti-Goat IgM (Mu chain) Secondary Antibody, HRP

Catalog No. PA129384
Change view
Click to view available options
Quantité:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantité
PA129384 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PA129384 Supplier Invitrogen™ Supplier No. PA129384
  • ONLINE EXCLUSIVE PRICE
    $566.34
    $329.25 / chaque

    Économisez $237.09

    42.0 % de rabais Les écarts et divulgations suivants peuvent entraîner des prix inférieurs aux rabais indiqués, sans toutefois entraîner de réductions de prix supplémentaires : Le prix affiché reflète une réduction promotionnelle temporaire. Aucun achat additionnel ni engagement commercial n’est requis pour bénéficier du prix réduit. Le prix peut être modifié en tout temps. Le prix promotionnel est valide uniquement pour les commandes en ligne. Votre tarification contractuelle peut différer. D’autres exclusions peuvent s’appliquer.

missing translation for 'validMainframeMsg'

Rabbit Polyclonal Secondary Antibody

PA1-29384 detects Goat IgM mu chain specific from goat samples.

Anti-Goat secondary antibodies are affinity-purified antibodies with well-characterized specificity for goat immunoglobulins and are useful in the detection, sorting or purification of its specified target. Secondary antibodies offer increased versatility enabling users to use many detection systems (e.g. HRP, AP, fluorescence). They can also provide greater sensitivity through signal amplification as multiple secondary antibodies can bind to a single primary antibody. Most commonly, secondary antibodies are generated by immunizing the host animal with a pooled population of immunoglobulins from the target species and can be further purified and modified (i.e. immunoaffinity chromatography, antibody fragmentation, label conjugation, etc.) to generate highly specific reagents.

Specifications

Antigène Goat IgM (Mu chain)
Applications ELISA, Immunohistochemistry, Western Blot
Classification Polyclonal
Concentration 10 mg/mL
Conjugué HRP
Formule PBS with 1% BSA and 0.01% gentamicin sulfate; pH 7.2
Espèces hôtes Rabbit
Immunogène Goat IgM mu heavy chain.
Méthode de purification IgG fraction
Quantité 1 mg
Statut réglementaire RUO
Primaire ou secondaire Secondary
Espèces cibles Goat
Contenu et stockage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Type de produit Secondary Antibody
Forme Liquid
Isotype IgG
Show More Show Less