Learn More
Rabbit anti-Goat IgG (H+L), Superclonal™ Recombinant Secondary Antibody, Alexa Fluor™ 555, Invitrogen™
Numéro de catalogue. A27017
Encompass
Numéro de catalogue. A27017 Fournisseur Thermo Fisher Scientific Code fournisseur LSA27017
Rabbit Recombinant Polyclonal Secondary Antibody
The sensitivity and specificity of each lot is confirmed using ELISA. Minimal cross-reactivity with mouse, rat, human, bovine, guinea pig, and donkey IgG is observed. Recombinant rabbit polyclonal antibodies are unique offerings from Thermo Fisher Scientific. They are comprised of a selection of multiple different recombinant monoclonal antibodies, providing the best of both worlds – the sensitivity of polyclonal antibodies with the specificity of monoclonal antibodies - all delivered with the consistency only found in a recombinant antibody. While functionally the same as a polyclonal antibody – recognizing multiple epitope sites on the target and producing higher detection sensitivity for low abundance targets – a recombinant rabbit polyclonal antibody has a known mixture of light and heavy chains. The exact population can be produced in every lot, circumventing the biological variability typically associated with polyclonal antibody production.
Goat IgG (H+L) Recombinant Polyclonal antibody specifically detects Goat IgG (H+L) in Goat samples. It is validated for Immunocytochemistry.