missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Rabbit anti-Goat IgG (H+L) Cross-Adsorbed Secondary Antibody, Biotin-XX

Catalog No. A10518
Encompass
Change view
Click to view available options
Quantity:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A10518 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A10518 Supplier Invitrogen™ Supplier No. A10518

missing translation for 'validMainframeMsg'

Rabbit Polyclonal Secondary Antibody

Rabbit anti-Goat IgG (H+L) Cross-Adsorbed Secondary Antibody for ICC/IF

Anti-Goat secondary antibodies are affinity-purified antibodies with well-characterized specificity for goat immunoglobulins and are useful in the detection, sorting or purification of its specified target. Secondary antibodies offer increased versatility enabling users to use many detection systems (e.g. HRP, AP, fluorescence). They can also provide greater sensitivity through signal amplification as multiple secondary antibodies can bind to a single primary antibody. Most commonly, secondary antibodies are generated by immunizing the host animal with a pooled population of immunoglobulins from the target species and can be further purified and modified (i.e. immunoaffinity chromatography, antibody fragmentation, label conjugation, etc.) to generate highly specific reagents.

Specifications

Antigen Goat IgG (H+L) Cross-Adsorbed
Applications Immunocytochemistry
Classification Polyclonal
Concentration 2 mg/mL
Conjugate Biotin-XX
Formulation PBS with 5mM sodium azide; pH 7.5
Host Species Rabbit
Immunogen Gamma Immunoglobins Heavy and Light chains.
Purification Method Purified
Quantity 1 mg
Regulatory Status RUO
Primary or Secondary Secondary
Target Species Goat
Content And Storage 4°C
Product Type Secondary Antibody
Form Liquid
Isotype IgG
Show More Show Less