missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IgG Rabbit anti-Goat, FITC, Polyclonal, Secondary Antibody, MilliporeSigma™
Description
Detect Goat IgG using this Rabbit anti-Goat IgG Antibody, FITC conjugate validated for use in immunofluoresence.
Spécifications
Spécifications
| Antigène | Rabbit anti-Goat IgG |
| Applications | Immunofluorescence |
| Classification | Polyclonal |
| Conjugué | FITC |
| Formule | Liquid in 0.01M Phosphate buffer, 0.25M NaCl pH 7.6 with 15mg/mL BSA, 0.01% Thimerosal and 0.05% sodium azide. RECONSTITUTION: Reconstitute to 1mg/mL with sterile distilled water. |
| Espèces hôtes | Rabbit |
| Immunogène | Purified Goat IgG. |
| Quantité | 1.5 mg |
| Primaire ou secondaire | Secondary |
| Espèces cibles | Goat |
| Afficher plus de résultats |
For Research Use Only