missing translation for 'onlineSavingsMsg'
Learn More

IgG Rabbit anti-Goat, HRP, Polyclonal, Secondary Antibody, MilliporeSigma™

Numéro de catalogue. AP107PMI
Modifier l'affichage
Click to view available options
Quantité:
1.5 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantité
AP107PMI 1.5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Numéro de catalogue. AP107PMI Fournisseur MilliporeSigma Code fournisseur AP107P

Pour de l'aide SVP utiliser la function de clavardage; nous envoyer un couriel à help@thermofisher.com ou appelez le service à la clientèle au 1-800-234-7437.

Rabbit Polyclonal Antibody

Rabbit anti-Goat IgG Antibody, flow cytometry, HRP conjugate is an antibody against Goat IgG for use in ELISA and western blotting.

Spécifications

Antigène Rabbit anti-Goat IgG. Fc
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 1.0mg/mL
Conjugué HRP
Formule Lyophilized. Buffer = 0.01M Sodium Phosphate, 0.25M NaCl, pH 7.6, with 15mg/mL BSA and 0.01% Thimerosal. RECONSTITUTION: Reconstitute with sterile distilled water to match the volume indicated on the label. Centrifuge product if it is not completely
Espèces hôtes Rabbit
Quantité 1.5 mL
Primaire ou secondaire Secondary
Espèces cibles Goat
Contenu et stockage 2-8°C lyophilized for up to 12 months −20°C in undiluted aliquots for up to 12 months. Avoid repeated freeze/thaw cycles.
Forme Lyophilized
Afficher plus de résultats Afficher moins de résultats

For Research Use Only