missing translation for 'onlineSavingsMsg'
Learn More

IgG Rabbit anti-Goat, HRP, Polyclonal, Secondary Antibody, MilliporeSigma™

Catalog No. AQ106PMI
Change view
Click to view available options
Quantity:
500 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AQ106PMI 500 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AQ106PMI Supplier MilliporeSigma Supplier No. AQ106P

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Rabbit Polyclonal Antibody

Detect Goat IgG using this Rabbit anti-Goat IgG Antibody, F(ab′) 2, HRP conjugate validated for use in ELISA and western blotting.

Specifications

Antigen Rabbit anti-Goat IgG F(ab')2
Applications ELISA, Western Blot
Classification Polyclonal
Conjugate HRP
Host Species Rabbit
Quantity 500 μL
Primary or Secondary Secondary
Target Species Goat

For Research Use Only