missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Rabbit anti-Feline IgG Fc Secondary Antibody [Janelia Fluor 669], R&D Systems™
Description
IgG Fc Polyclonal antibody specifically detects IgG Fc in Feline samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin), Dot Blot
Spécifications
Spécifications
| Antigène | IgG Fc |
| Applications | Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin), Dot Blot |
| Classification | Polyclonal |
| Conjugué | Janelia Fluor 669 |
| Formule | 50mM Sodium Borate |
| Espèces hôtes | Rabbit |
| Immunogène | Cat IgG F(c) fragment |
| Méthode de purification | Multi-step |
| Quantité | 0.5 mL |
| Statut réglementaire | RUO |
| Afficher plus de résultats |