missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IgG Rabbit anti-Chicken, Alkaline Phosphatase, Polyclonal, Secondary Antibody, MilliporeSigma™
Description
Rabbit anti-Chicken IgG Antibody, Alkaline Phosphatase conjugate detects level of Chicken IgG and has been published and validated for use in ELISA and western blotting.
Spécifications
Spécifications
| Antigène | Rabbit anti-Chicken IgG |
| Applications | ELISA, Western Blot |
| Classification | Polyclonal |
| Conjugué | Alkaline Phosphatase (AP) |
| Espèces hôtes | Rabbit |
| Quantité | 1 mL |
| Primaire ou secondaire | Secondary |
| Espèces cibles | Chicken |
For Research Use Only