missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(R)-(+)-TolBINAP 98,0+%, TCI America™
Spécifications
Spécifications
| Melting Point | 258°C |
| Color | Blanc-jaune |
| Quantity | 1 g |
| Formula Weight | 678.80 |
| Percent Purity | ≥98.0% (HPLC) |
| Physical Form | Poudre cristalline |
| Chemical Name or Material | (R)-(+)-TolBINAP |
Sécurité et manipulation
| TSCA | No |