missing translation for 'onlineSavingsMsg'
Learn More

(R,R)-(-)-N,N'-Bis(3,5-di-tert-butylsalicylidene)-1,2-cyclohexanediaminomanganese(III) Chloride 98.0+%, TCI America™

Catalog No. B56135G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B56135G 5 g
B56131G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B56135G Supplier TCI America Supplier No. B56135G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 138124-32-0
Molecular Formula C36H54ClMnN2O2-
Molecular Weight (g/mol) 637.228
MDL Number MFCD02101664
InChI Key YRVXCOIDMFNGIJ-SEILFYAJSA-M
Synonym 1r,2r---1,2-cyclohexanediamino-n,n'-bis 3,5-di-t-butylsalicylidene manganese iii chloride r,r-jacobsen cat.
PubChem CID 131675872
IUPAC Name 2,4-ditert-butyl-6-[[[(1R,2R)-2-[(3,5-ditert-butyl-6-oxocyclohexa-2,4-dien-1-ylidene)methylamino]cyclohexyl]amino]methylidene]cyclohexa-2,4-dien-1-one;manganese;chloride
SMILES CC(C)(C)C1=CC(=CNC2CCCCC2NC=C3C=C(C=C(C3=O)C(C)(C)C)C(C)(C)C)C(=O)C(=C1)C(C)(C)C.[Cl-].[Mn]

Specifications

Melting Point 326°C
Quantity 5 g
Formula Weight 635.21
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material (R,R)-(-)-N,N′-Bis(3,5-di-tert-butylsalicylidene)-1,2-cyclohexanediaminomanganese(III) Chlorid