missing translation for 'onlineSavingsMsg'
Learn More

(R,R)-(-)-2,3-Dimethoxy-1,4-bis(dimethylamino)butane 90.0+%, TCI America™

Numéro de catalogue. D23955G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantity
D23955G 5 g
D23951G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Numéro de catalogue. D23955G Fournisseur TCI America Code fournisseur D23955G
Il en reste null

Identifiants chimiques

CAS 26549-22-4
Molecular Formula C10H24N2O2
Molecular Weight (g/mol) 204.314
MDL Number MFCD00008331
InChI Key VYQCQNCBTMHEFI-NXEZZACHSA-N
PubChem CID 2724116
IUPAC Name (2R,3R)-2,3-dimethoxy-N,N,N',N'-tetramethylbutane-1,4-diamine
SMILES CN(C)CC(C(CN(C)C)OC)OC

Spécifications

Color Yellow
Boiling Point 64°C
Quantity 5 g
Synonym (R,R)-(-)-DDB, (R,R)-(-)-2,3-Dimethoxy-N,N,N′,N′-tetramethyl-1,4-butanediamine
Formula Weight 204.31
Percent Purity ≥90.0% (GC)
Physical Form Liquid
Chemical Name or Material (R,R)-(-)-2,3-Dimethoxy-1,4-bis(dimethylamino)butane