missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(R,R)-(-)-2,3-Dimethoxy-1,4-bis(dimethylamino)butane 90.0+%, TCI America™
Spécifications
Spécifications
| Color | Yellow |
| Boiling Point | 64°C |
| Quantity | 1 g |
| Synonym | (R,R)-(-)-DDB, (R,R)-(-)-2,3-Dimethoxy-N,N,N′C,N′C-tetramethyl-1,4-butanediamine |
| Formula Weight | 204.31 |
| Percent Purity | ≥90.0% (GC) |
| Physical Form | Liquid |
| Chemical Name or Material | (R,R)-(-)-2,3-Dimethoxy-1,4-bis(dimethylamino)butane |
Sécurité et manipulation
| TSCA | No |