missing translation for 'onlineSavingsMsg'
Learn More

(R)-(-)-Prolinol, 98+%

Catalog No. AAL1110903
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1110903 1 g
AAL1110906 5 g
AAL1110914 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAL1110903 Supplier Thermo Scientific Chemicals Supplier No. L1110903

CAS: 68832-13-3 | C5H11NO | 101.149 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 68832-13-3
Molecular Formula C5H11NO
Molecular Weight (g/mol) 101.149
MDL Number MFCD00064321
InChI Key HVVNJUAVDAZWCB-RXMQYKEDSA-N
Synonym d-prolinol, r-pyrrolidin-2-ylmethanol, 2r-pyrrolidin-2-ylmethanol, r---2-pyrrolidinemethanol, r-prolinol, 2r-pyrrolidin-2-yl methanol, r-1-pyrrolidin-2-yl-methanol, 2-pyrrolidinemethanol, 2r, r---2-hydroxymethyl pyrrolidine
PubChem CID 2724541
ChEBI CHEBI:84258
IUPAC Name [(2R)-pyrrolidin-2-yl]methanol
SMILES C1CC(NC1)CO

Specifications

Refractive Index 1.4849
Density 1.025
Boiling Point 74°C to 76°C (2mmHg)
Quantity 1 g
Flash Point 86°C (186°F)
Optical Rotation −31° (c=1 in Toluene)
Sensitivity Air sensitive; Hygroscopic
Beilstein 1523669
Formula Weight 101.15
Percent Purity ≥98%
Chemical Name or Material (R)-(-)-Prolinol
Show More Show Less
Hazard Category H227-H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H227
Causes skin irritation.
Causes serious eye irritation.
Combustible liquid.
Precautionary Statement P210-P235-P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P370+P378q-P501c
EINECSNumber 629-147-4
TSCA No
Recommended Storage Ambient temperatures; Store under Nitrogen

RUO – Research Use Only