missing translation for 'onlineSavingsMsg'
Learn More

(R)-(-)-Phenyl-1,2-ethanediol, 99%, Thermo Scientific Chemicals

Catalog No. AAL1049403
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1049403 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAL1049403 Supplier Thermo Scientific Chemicals Supplier No. L1049403
Get Up to $1,800 in Chemicals with Purchase
View all promotions

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 16355-00-3
Molecular Formula C8H10O2
Molecular Weight (g/mol) 138.166
MDL Number MFCD00064262
InChI Key PWMWNFMRSKOCEY-QMMMGPOBSA-N
Synonym r---1-phenyl-1,2-ethanediol, r-1-phenylethane-1,2-diol, 1r-1-phenylethane-1,2-diol, r---1-phenylethane-1,2-diol, r-1-phenyl-1,2-ethanediol, styrene glycol,-, unii-2lzm4b71kf, r-+-1-phenylethane-1,2-diol, 2lzm4b71kf
PubChem CID 2724621
IUPAC Name (1R)-1-phenylethane-1,2-diol
SMILES C1=CC=C(C=C1)C(CO)O

Specifications

Melting Point 67°C to 69°C
Boiling Point 272°C to 274°C
Quantity 1 g
Beilstein 2355547
Optical Rotation −39° (c=3 in Ethanol)
Formula Weight 138.17
Percent Purity 99%
Chemical Name or Material (R)-(-)-Phenyl-1,2-ethanediol
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only