missing translation for 'onlineSavingsMsg'
Learn More

(R)-(-)-NBD-APy 98.0+%, TCI America™

Catalog No. A5563100MG
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A5563100MG 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A5563100MG Supplier TCI America Supplier No. A5563100MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

[HPLC Labeling Reagent for e.e.Determination]

Chemical Identifiers

CAS 143112-51-0
Molecular Formula C10H11N5O3
Molecular Weight (g/mol) 249.23
MDL Number MFCD00191510
InChI Key QSDDQXROWUJAJX-ZCFIWIBFSA-N
Synonym (R)-(-)-4-Nitro-7-(3-aminopyrrolidin-1-yl)benzofurazan, (R)-(-)-4-Nitro-7-(3-aminopyrrolidin-1-yl)-2,1,3-benzoxadiazole
PubChem CID 15682840
IUPAC Name (3R)-1-(4-nitro-2,1,3-benzoxadiazol-7-yl)pyrrolidin-3-amine
SMILES C1CN(CC1N)C2=CC=C(C3=NON=C23)[N+](=O)[O-]

Specifications

Quantity 100 mg
Formula Weight 249.23
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material (R)-(-)-NBD-APy [=(R)-(-)-4-Nitro-7-(3-aminopyrrolidin-1-yl)-2,1,3-benzoxadiazole] [HPLC Labe