missing translation for 'onlineSavingsMsg'
Learn More

(R)-(-)-Linalool, 95% (sum of enantiomers)

Catalog No. AC428311000
Change view
Click to view available options
Quantity:
25 g
100 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC428311000 100 g Glass bottle
AC428310250 25 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC428311000 Supplier Thermo Scientific Chemicals Supplier No. 428311000
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 126-91-0 | C10H18O | 154.25 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 126-91-0
Molecular Formula C10H18O
Molecular Weight (g/mol) 154.25
MDL Number MFCD00135469
InChI Key CDOSHBSSFJOMGT-JTQLQIEISA-N
Synonym --linalool, 3r-linalool, r---linalool, unii-3u21e3v8i2, 1,6-octadien-3-ol, 3,7-dimethyl-, 3r, 3r-3,7-dimethylocta-1,6-dien-3-ol, l-linalool, 3r,7-dimethylocta-1,6-dien-3-ol, r-3,7-dimethylocta-1,6-dien-3-ol, linalool +
PubChem CID 443158
ChEBI CHEBI:28
IUPAC Name (3R)-3,7-dimethylocta-1,6-dien-3-ol
SMILES CC(=CCCC(C)(C=C)O)C

Specifications

Color Colorless to Yellow
Density 0.8620g/mL
Boiling Point 196.0°C to 198.0°C
Flash Point 75°C
Infrared Spectrum Authentic
Packaging Glass bottle
Refractive Index 1.4600 to 1.4640 (20°C, 589nm)
Quantity 100 g
Specific Gravity 0.862
Merck Index 15, 5550
Specific Rotation -15° to -21° (20°C, 589nm) (neat)
Formula Weight 154.25
Percent Purity 95% (sum of enantiomers)
Physical Form Liquid
Chemical Name or Material (R)-(-)-Linalool, 95% (sum of enantiomers)
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Causes skin irritation.
May cause an allergic skin reaction.
Precautionary Statement GHS P Statement
IF ON SKIN: Wash with plenty of soap and water.
If skin irritation occurs: Get medical advice/attention.
If skin irritation or rash occurs: Get medical advice/attention.
EINECSNumber 204-811-2

RUO – Research Use Only