missing translation for 'onlineSavingsMsg'
Learn More

(R)-(+)-Limonene, 96%

Catalog No. AAL04733AE
Change view
Click to view available options
Quantity:
100 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL04733AE 100 mL
AAL04733AP 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL04733AE Supplier Thermo Scientific Chemicals Supplier No. L04733AE
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 5989-27-5 | C10H16 | 136.24 g/mol

Chiral building block. Biodegradable solvent. Limonene is used as a solvent for filament-fused 3D printing, in cosmetic products, as a fragrance and as a biofuel. It is useful to promote weight loss, prevent cancer, treat cancer and treat bronchitis. It also finds use in foods, beverages, chewing gum, as a flavoring agent and a shrinking agent to dissolve polystyrene. It acts as a chiral intermediate for natural product synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Limonene is used as a solvent for filament-fused 3D printing, in cosmetic products, as a fragrance and as a biofuel. It is useful to promote weight loss, prevent cancer, treat cancer and treat bronchitis. It also finds use in foods, beverages, chewing gum, as a flavoring agent and a shrinking agent to dissolve polystyrene. It acts as a chiral intermediate for natural product synthesis and as an insect repellant.

Solubility
Miscible with alcohol,chloroform,ether and carbondisulfide.Immiscible with water.

Notes
Incompatible with strong oxidizing agents. Store in a cool place.

Chemical Identifiers

CAS 5989-27-5
Molecular Formula C10H16
Molecular Weight (g/mol) 136.24
MDL Number MFCD00062991
InChI Key XMGQYMWWDOXHJM-JTQLQIEISA-N
Synonym d-limonene, +-limonene, r-+-limonene, +-4r-limonene, +-carvene, +-dipentene, citrene, +-p-mentha-1,8-diene, 4r-limonene, d-+-limonene
PubChem CID 440917
ChEBI CHEBI:15382
IUPAC Name (4R)-1-methyl-4-prop-1-en-2-ylcyclohexene
SMILES CC(=C)[C@@H]1CCC(C)=CC1

Specifications

Melting Point -74°C
Density 0.841
Boiling Point 177°C to 178°C
Flash Point 48°C (118°F)
Odor Fruit-like
Refractive Index 1.473
Quantity 100 mL
UN Number UN2052
Beilstein 2204754
Merck Index 14,5493
Solubility Information Miscible with alcohol,chloroform,ether and carbondisulfide.Immiscible with water.
Optical Rotation +115° (c=10 in Ethanol)
Formula Weight 136.23
Percent Purity 96%
Chemical Name or Material (R)-(+)-Limonene, stabilized
Show More Show Less
Hazard Category H226-H304-H315-H317-H319
Hazard Statement GHS H Statement
H226-H315-H317
Flammable liquid and vapor.
Causes skin irritation.
May cause an allergic skin reaction.
Precautionary Statement P210-P233-P240-P241-P242-P243-P261-P264b-P272-P280-P301+P310-P303+P361+P353-P305+P351+P338-P331-P333+P313-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: DIPENTENE
EINECSNumber 227-813-5
RTECSNumber GW6360000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only