missing translation for 'onlineSavingsMsg'
Learn More

(R)-Glycidyl 3-Nitrobenzenesulfonate 98.0+%, TCI America™

Catalog No. G02865G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
G02865G 5 g
G028625G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. G02865G Supplier TCI America Supplier No. G02865G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 115314-17-5
Molecular Formula C9H9NO6S
Molecular Weight (g/mol) 259.232
MDL Number MFCD00013436
InChI Key AIHIHVZYAAMDPM-MRVPVSSYSA-N
Synonym r---glycidyl nosylate, r-glycidyl nosylate, r-oxiran-2-ylmethyl 3-nitrobenzenesulfonate, r-glycidyl 3-nitrobenzenesulfonate, r---glycidyl 3-nitrobenzenesulfonate, r---glycidyl-3-nitrobenzenesulfonate, ccris 6393, oxiranylmethyl r-3-nitrobenzenesulfonate, benzenesulfonic acid, 3-nitro-, oxiranylmethyl ester, r
PubChem CID 146513
IUPAC Name [(2R)-oxiran-2-yl]methyl 3-nitrobenzenesulfonate
SMILES C1C(O1)COS(=O)(=O)C2=CC=CC(=C2)[N+](=O)[O-]

Specifications

Melting Point 63°C
Color White-Yellow
Quantity 5 g
UN Number 1325
Formula Weight 259.23
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material (R)-Glycidyl 3-Nitrobenzenesulfonate
RTECSNumber DB7192000
TSCA No