missing translation for 'onlineSavingsMsg'
Learn More

(R)-(+)-alpha,alpha-Diphenylprolinol, 99%, Thermo Scientific™

Catalog No. AAL0921803
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0921803 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAL0921803 Supplier Thermo Scientific Chemicals Supplier No. L0921803
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Chemical Identifiers

CAS 22348-32-9
Molecular Formula C17H19NO
Molecular Weight (g/mol) 253.345
MDL Number MFCD00077754
InChI Key OGCGXUGBDJGFFY-MRXNPFEDSA-N
Synonym r-diphenyl pyrrolidin-2-yl methanol, r-+-alpha,alpha-diphenyl-2-pyrrolidinemethanol, r-+-2-diphenylhydroxymethyl pyrrolidine, diphenyl 2r-pyrrolidin-2-ylmethanol, r-+-2-diphenylhydroxymethyl-pyrrolidin, diphenyl-2r-pyrrolidin-2-yl methanol, r-alpha,alpha-diphenyl-2-pyrrolidinemethanol, alpha,alpha-diphenyl-d-prolinol, r-+-alpha,alpha-diphenylprolinol, diphenyl 2r-pyrrolidin-2-yl methanol
PubChem CID 7045371
IUPAC Name diphenyl-[(2R)-pyrrolidin-2-yl]methanol
SMILES C1CC(NC1)C(C2=CC=CC=C2)(C3=CC=CC=C3)O

Specifications

Melting Point 76°C to 78°C
Quantity 1 g
Optical Rotation +57.5° (c=3 in Methanol)
Beilstein 4353363
Formula Weight 253.35
Percent Purity 99%
Chemical Name or Material (R)-(+)-alpha,alpha-Diphenylprolinol
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only