missing translation for 'onlineSavingsMsg'
Learn More

(R)-(-)-4-Phenyl-2-oxazolidinone 99.0+%, TCI America™

Catalog No. P13075G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P13075G 5 g
P130725G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P13075G Supplier TCI America Supplier No. P13075G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 90319-52-1
Molecular Formula C9H9NO2
Molecular Weight (g/mol) 163.176
MDL Number MFCD00192393
InChI Key QDMNNMIOWVJVLY-QMMMGPOBSA-N
Synonym r---4-phenyl-2-oxazolidinone, r-4-phenyloxazolidin-2-one, r-4-phenyl-2-oxazolidinone, 4r-4-phenyl-1,3-oxazolidin-2-one, 4r-4-phenyloxazolidin-2-one, 2-oxazolidinone, 4-phenyl-, 4r, r-4-phenyl-oxazolidin-2-one, pubchem6077, ksc131o9d, 4 r-phenyl-2-oxazolidinone
PubChem CID 730425
IUPAC Name (4R)-4-phenyl-1,3-oxazolidin-2-one
SMILES C1C(NC(=O)O1)C2=CC=CC=C2

Specifications

Melting Point 131°C
Color White
Quantity 5 g
Formula Weight 163.18
Percent Purity ≥99.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material (R)-(-)-4-Phenyl-2-oxazolidinone