missing translation for 'onlineSavingsMsg'
Learn More

(R)-(-)-4-Phenyl-2-oxazolidinone, 98%

Catalog No. AAH2717106
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH2717106 5 g
AAH2717103 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH2717106 Supplier Thermo Scientific Chemicals Supplier No. H2717106

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 90319-52-1 | C9H9NO2 | 163.176 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 90319-52-1
Molecular Formula C9H9NO2
Molecular Weight (g/mol) 163.176
MDL Number MFCD00192393
InChI Key QDMNNMIOWVJVLY-QMMMGPOBSA-N
Synonym r---4-phenyl-2-oxazolidinone, r-4-phenyloxazolidin-2-one, r-4-phenyl-2-oxazolidinone, 4r-4-phenyl-1,3-oxazolidin-2-one, 4r-4-phenyloxazolidin-2-one, 2-oxazolidinone, 4-phenyl-, 4r, r-4-phenyl-oxazolidin-2-one, pubchem6077, ksc131o9d, 4 r-phenyl-2-oxazolidinone
PubChem CID 730425
IUPAC Name (4R)-4-phenyl-1,3-oxazolidin-2-one
SMILES C1C(NC(=O)O1)C2=CC=CC=C2

Specifications

Melting Point 129°C to 132°C
Quantity 5 g
Optical Rotation −55° (c=2 in Chloroform)
Formula Weight 163.18
Percent Purity 98%
Chemical Name or Material (R)-(-)-4-Phenyl-2-oxazolidinone
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only