missing translation for 'onlineSavingsMsg'
Learn More

(R)-(-)-4-Fluorostyrene oxide, 98+%, ee 98+%, Thermo Scientific Chemicals

Catalog No. AAH3360306
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH3360306 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAH3360306 Supplier Thermo Scientific Chemicals Supplier No. H3360306
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 134356-73-3 | Molecular Formula: C8H7FO | Molecular Weight: 138.14 g/mol

It is used as pharmaceutical intermediate. Benzoylthiophenes are allosteric enhancers (AE) of agonist activity at the A1 adenosine receptor.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used as pharmaceutical intermediate. Benzoylthiophenes are allosteric enhancers (AE) of agonist activity at the A1 adenosine receptor.

Solubility
Sparingly soluble in water.(0.26 g/L) (25°C),

Notes
Store in cool dry place. Ensure proper ventilation. Incompatible with oxidizing agents.

Chemical Identifiers

CAS 134356-73-3
Molecular Formula C8H7FO
Molecular Weight (g/mol) 138.14
MDL Number MFCD03788740,MFCD00043994,MFCD03788740
InChI Key ICVNPQMUUHPPOK-UHFFFAOYNA-N
Synonym r-4-fluorophenyl oxirane, r-2-4-fluorophenyl oxirane, 2r-2-4-fluorophenyl oxirane, r-4-fluorostyrene oxide, r---4-fluorostyrene oxide, pubchem18166, r-4-fluorophenyl oxirane, technical gc
PubChem CID 7023123
IUPAC Name (2R)-2-(4-fluorophenyl)oxirane
SMILES FC1=CC=C(C=C1)C1CO1

Specifications

Density 1.17
Boiling Point 92°C (14 mmHg)
Refractive Index 1.5087
Quantity 5 g
UN Number UN1992
Solubility Information Sparingly soluble in water.(0.26g/L) (25°C),
Formula Weight 138.14
Percent Purity ≥98%
Assay ee ≥98%
Chemical Name or Material (R)-(-)-4-Fluorostyrene oxide
Hazard Category H226-H302+H312+H332-H315-H319-H335-H351
Hazard Statement GHS H Statement
H331-H226-H351-H302-H312-H315-H319-H335
Toxic if inhaled.
Flammable liquid and vapor.
Suspected of causing cancer.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P201-P202-P210-P233-P235-P240-P241-P242-P243-P261-P264b-P270-P271-P280i-P281-P301+P312-P303+P361+P353-P304+P340-P305+P351+P338-P308+P313-P312-P330-P332+P313-P363-P370+P378q-P5
DOTInformation Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, TOXIC, N.O.S.
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only