missing translation for 'onlineSavingsMsg'
Learn More

(R)-(+)-3-Hydroxy-3-phenylpropionic acid, 98+%

Catalog No. AAL19040MD
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL19040MD 250 mg
AAL1904003 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL19040MD Supplier Thermo Scientific Chemicals Supplier No. L19040MD
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 2768-42-5 | C9H9O3 | 165.17 g/mol

It is employed in the synthesis and evaluation of a novel, reusable solid-supported open chain chiral auxiliary derived from m-hydrobenzoin for the diastereoselective reduction of α-keto esters.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is employed in the synthesis and evaluation of a novel, reusable solid-supported open chain chiral auxiliary derived from m-hydrobenzoin for the diastereoselective reduction of α-keto esters.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.

Chemical Identifiers

CAS 2768-42-5
Molecular Formula C9H9O3
Molecular Weight (g/mol) 165.17
MDL Number MFCD00145219
InChI Key AYOLELPCNDVZKZ-MRVPVSSYSA-M
Synonym 3r-3-hydroxy-3-phenylpropanoic acid, r-3-hydroxy-3-phenylpropanoic acid, r-3-hydroxy-3-phenylpropionic acid, r-+-3-hydroxy-3-phenylpropionic acid, +-3-phenylhydracrylic acid, benzenepropanoic acid, b-hydroxy-, br, r-+-3-hydroxy-3-phenylpropanoic acid, benzenepropanoic acid,, a-hydroxy-,, ar, r-+-3-hydroxy-3-phenyl propionic acid, unii-s2w4lws09c component
PubChem CID 6950815
ChEBI CHEBI:51059
IUPAC Name (3R)-3-hydroxy-3-phenylpropanoic acid
SMILES O[C@H](CC([O-])=O)C1=CC=CC=C1

Specifications

Melting Point 118°C to 121°C
Quantity 250 mg
Beilstein 3197772
Optical Rotation +22° (c=4 in Methanol)
Formula Weight 166.18
Percent Purity ≥98%
Chemical Name or Material (R)-(+)-3-Hydroxy-3-phenylpropionic acid
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only