Learn More
(R)-3-(Boc-amino)piperidine, 97%
CAS: 309956-78-3 | C10H20N2O2 | 200.282 g/mol
$98.37 - $663.30
Chemical Identifiers
| CAS | 309956-78-3 |
|---|---|
| Molecular Formula | C10H20N2O2 |
| Molecular Weight (g/mol) | 200.282 |
| MDL Number | MFCD03093382 |
| InChI Key | WUOQXNWMYLFAHT-MRVPVSSYSA-N |
| Synonym | r-3-boc-aminopiperidine, r-3-boc-amino piperidine, r-tert-butyl piperidin-3-ylcarbamate, tert-butyl n-3r-piperidin-3-yl carbamate, tert-butyl r-piperidin-3-ylcarbamate, r-3-tert-butoxycarbonylamino piperidine, r-3-boc-amino-piperidine, r-+-3-tert-butoxycarbonylaminopiperidine, r-piperidin-3-yl-carbamic acid tert-butyl ester |
| PubChem CID | 1514172 |
| IUPAC Name | tert-butyl N-[(3R)-piperidin-3-yl]carbamate |
| SMILES | CC(C)(C)OC(=O)NC1CCCNC1 |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|
AAH5274903
|
N/A | 1 g |
Each for $98.37
|
|
|||||
|
AAH5274906
|
N/A | 5 g |
Each for $336.00
|
|
|||||
|
AAH5274914
|
N/A | 25 g |
Each for $663.30
|
|
|||||
Description
(R)-3-(Boc-amino)piperidine is used as an organic chemical synthesis intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications(R)-3-(Boc-amino)piperidine is used as an organic chemical synthesis intermediate.
Solubility
Soluble in methanol and ethanol.
Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
Chemical Identifiers
| 309956-78-3 | |
| 200.282 | |
| WUOQXNWMYLFAHT-MRVPVSSYSA-N | |
| 1514172 | |
| CC(C)(C)OC(=O)NC1CCCNC1 |
| C10H20N2O2 | |
| MFCD03093382 | |
| r-3-boc-aminopiperidine, r-3-boc-amino piperidine, r-tert-butyl piperidin-3-ylcarbamate, tert-butyl n-3r-piperidin-3-yl carbamate, tert-butyl r-piperidin-3-ylcarbamate, r-3-tert-butoxycarbonylamino piperidine, r-3-boc-amino-piperidine, r-+-3-tert-butoxycarbonylaminopiperidine, r-piperidin-3-yl-carbamic acid tert-butyl ester | |
| tert-butyl N-[(3R)-piperidin-3-yl]carbamate |
Specifications
| 309956-78-3 | |
| MFCD03093382 | |
| Soluble in methanol and ethanol. | |
| CC(C)(C)OC(=O)NC1CCCNC1 | |
| 200.282 | |
| 200.28 | |
| (R)-3-(Boc-amino)piperidine |
| C10H20N2O2 | |
| r-3-boc-aminopiperidine, r-3-boc-amino piperidine, r-tert-butyl piperidin-3-ylcarbamate, tert-butyl n-3r-piperidin-3-yl carbamate, tert-butyl r-piperidin-3-ylcarbamate, r-3-tert-butoxycarbonylamino piperidine, r-3-boc-amino-piperidine, r-+-3-tert-butoxycarbonylaminopiperidine, r-piperidin-3-yl-carbamic acid tert-butyl ester | |
| WUOQXNWMYLFAHT-MRVPVSSYSA-N | |
| tert-butyl N-[(3R)-piperidin-3-yl]carbamate | |
| 1514172 | |
| 97% |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only