accessibility menu, dialog, popup

UserName

H3CH3CCH3OONH(R)HN

CAS RN 309956-78-3

IUPAC Name: tert-butyl N-[(3R)-piperidin-3-yl]carbamate
Synonyms: tert-butyl N-[(3R)-piperidin-3-yl]carbamate
Molecular Weight (g/mol): 200.28
Molecular Formula: C10H20N2O2
InChi Key: WUOQXNWMYLFAHT-MRVPVSSYSA-N
SMILES: CC(C)(C)OC(=O)N[C@@H]1CCCNC1
Percent Purity 
  • (2)
  • (3)
Quantity 
  • (2)
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (3)
  • (2)
  • (3)
  • (2)
  • (1)
  • (2)