missing translation for 'onlineSavingsMsg'
Learn More

(R)-3-(Boc-amino)piperidine, 97%

CAS: 309956-78-3 | C10H20N2O2 | 200.282 g/mol

Supplier:  Thermo Scientific Chemicals H5274906

Quantity 5 g

 View more versions of this product

Catalog No. AAH5274906


Only null left

Description

Description

(R)-3-(Boc-amino)piperidine is used as an organic chemical synthesis intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
(R)-3-(Boc-amino)piperidine is used as an organic chemical synthesis intermediate.

Solubility
Soluble in methanol and ethanol.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
Specifications

Chemical Identifiers

309956-78-3
200.282
WUOQXNWMYLFAHT-MRVPVSSYSA-N
1514172
CC(C)(C)OC(=O)NC1CCCNC1
C10H20N2O2
MFCD03093382
r-3-boc-aminopiperidine, r-3-boc-amino piperidine, r-tert-butyl piperidin-3-ylcarbamate, tert-butyl n-3r-piperidin-3-yl carbamate, tert-butyl r-piperidin-3-ylcarbamate, r-3-tert-butoxycarbonylamino piperidine, r-3-boc-amino-piperidine, r-+-3-tert-butoxycarbonylaminopiperidine, r-piperidin-3-yl-carbamic acid tert-butyl ester
tert-butyl N-[(3R)-piperidin-3-yl]carbamate

Specifications

(R)-3-(Boc-amino)piperidine
C10H20N2O2
r-3-boc-aminopiperidine, r-3-boc-amino piperidine, r-tert-butyl piperidin-3-ylcarbamate, tert-butyl n-3r-piperidin-3-yl carbamate, tert-butyl r-piperidin-3-ylcarbamate, r-3-tert-butoxycarbonylamino piperidine, r-3-boc-amino-piperidine, r-+-3-tert-butoxycarbonylaminopiperidine, r-piperidin-3-yl-carbamic acid tert-butyl ester
WUOQXNWMYLFAHT-MRVPVSSYSA-N
tert-butyl N-[(3R)-piperidin-3-yl]carbamate
200.282
200.28
309956-78-3
MFCD03093382
Soluble in methanol and ethanol.
CC(C)(C)OC(=O)NC1CCCNC1
5 g
1514172
97%
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions