missing translation for 'onlineSavingsMsg'
Learn More

(R)-3-(Boc-amino)piperidine, 97%

Catalog No. AAH5274903
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH5274903 1 g
AAH5274906 5 g
AAH5274914 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAH5274903 Supplier Thermo Scientific Chemicals Supplier No. H5274903
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 309956-78-3 | C10H20N2O2 | 200.282 g/mol

(R)-3-(Boc-amino)piperidine is used as an organic chemical synthesis intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
(R)-3-(Boc-amino)piperidine is used as an organic chemical synthesis intermediate.

Solubility
Soluble in methanol and ethanol.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.

Chemical Identifiers

CAS 309956-78-3
Molecular Formula C10H20N2O2
Molecular Weight (g/mol) 200.282
MDL Number MFCD03093382
InChI Key WUOQXNWMYLFAHT-MRVPVSSYSA-N
Synonym r-3-boc-aminopiperidine, r-3-boc-amino piperidine, r-tert-butyl piperidin-3-ylcarbamate, tert-butyl n-3r-piperidin-3-yl carbamate, tert-butyl r-piperidin-3-ylcarbamate, r-3-tert-butoxycarbonylamino piperidine, r-3-boc-amino-piperidine, r-+-3-tert-butoxycarbonylaminopiperidine, r-piperidin-3-yl-carbamic acid tert-butyl ester
PubChem CID 1514172
IUPAC Name tert-butyl N-[(3R)-piperidin-3-yl]carbamate
SMILES CC(C)(C)OC(=O)NC1CCCNC1

Specifications

Solubility Information Soluble in methanol and ethanol.
Quantity 1 g
Formula Weight 200.28
Percent Purity 97%
Chemical Name or Material (R)-3-(Boc-amino)piperidine
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only