missing translation for 'onlineSavingsMsg'
Learn More

(R)-3-(Boc-amino)-3-(4-bromophenyl)propionic acid, 95%

Catalog No. AAH5216603
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH5216603 1 g
AAH5216606 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH5216603 Supplier Thermo Scientific Chemicals Supplier No. H5216603
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 261380-20-5 | C14H18BrNO4 | 344.205 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 261380-20-5
Molecular Formula C14H18BrNO4
Molecular Weight (g/mol) 344.205
MDL Number MFCD01320860
InChI Key ZAMLGGRVTAXBHI-LLVKDONJSA-N
Synonym boc-r-3-amino-3-4-bromo-phenyl-propionic acid, r-3-4-bromophenyl-3-tert-butoxycarbonyl amino propanoic acid, r-boc-4-bromo-beta-phe-oh, boc-4-bromo-l-beta-phenylalanine, boc-beta-r-4-bromophenylalanine, r-n-boc-3-amino-3-4-bromophenyl propanoic acid, boc-r-4-beta-bromophenylalanine, r-3-boc-amino-3-4-bromophenyl propionic acid, boc-r-4-bromo-beta-phenylalanine, r-n-boc-beta-4-bromophenyl-beta-alanine
PubChem CID 7009110
IUPAC Name (3R)-3-(4-bromophenyl)-3-[(2-methylpropan-2-yl)oxycarbonylamino]propanoic acid
SMILES CC(C)(C)OC(=O)NC(CC(=O)O)C1=CC=C(C=C1)Br

Specifications

Melting Point 143°C to 145°C
Quantity 1 g
Formula Weight 344.2
Percent Purity 95%
Chemical Name or Material (R)-3-(Boc-amino)-3-(4-bromophenyl)propionic acid
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only