missing translation for 'onlineSavingsMsg'
Learn More

(R)-(+)-3-Aminoquinuclidine dihydrochloride, 98%, Thermo Scientific™

Numéro de catalogue. AC382890010
Modifier l'affichage
Click to view available options
Quantité:
1g
Emballage:
Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantité Emballage
AC382890010 1g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Numéro de catalogue. AC382890010 Fournisseur Thermo Scientific Chemicals Code fournisseur 382890010

Pour de l'aide SVP utiliser la function de clavardage; nous envoyer un couriel à help@thermofisher.com ou appelez le service à la clientèle au 1-800-234-7437.

Pour de l'aide SVP utiliser la function de clavardage; nous envoyer un couriel à help@thermofisher.com ou appelez le service à la clientèle au 1-800-234-7437.

 

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Identifiants chimiques

CAS 123536-14-1
Formule moléculaire C7H14N2·2ClH
Poids moléculaire (g/mol) 199.12
Numéro MDL MFCD00191752
Clé InChI STZHBULOYDCZET-KLXURFKVSA-N
Synonyme r-3-aminoquinuclidine dihydrochloride, r-+-3-aminoquinuclidine dihydrochloride, r-quinuclidin-3-amine dihydrochloride, 3r-1-azabicyclo 2.2.2 octan-3-amine dihydrochloride, 3r-quinuclidin-3-amine dihydrochloride, r-+-3-aminoquinuclidine 2hcl, 3r-3-aminoquinuclidine dihydrochloride, r-1-aza-bicyclo 2.2.2 oct-3-ylamine dihydrochloride, 1-azabicyclo 2.2.2 octan-3-amine, dihydrochloride, 3r, pubchem9967
PubChem CID 12494972
Nom de l’IUPAC (3R)-1-azabicyclo[2.2.2]octan-3-amine;dihydrochloride
SOURIRES C1CN2CCC1C(C2)N.Cl.Cl

Spécifications

Nom ou substance chimique (R)-(+)-3-Aminoquinuclidine dihydrochloride
CAS % minimum 97.5
CAS % maximal 100.0
Spectre infrarouge Authentic
Fourchette de pourcentages d’essai 98%
Emballage Glass bottle
Quantité 1g
Rotation spécifique +22° to +26° (20°C, 589nm) (C=1, Water)
Poids de formule 199.12
Pourcentage de pureté 98%
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Harmful if swallowed.
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

RUO – Research Use Only