missing translation for 'onlineSavingsMsg'
Learn More

(R)-(+)-2-Methyl-CBS-oxazaborolidine, 1M solution in toluene, AcroSeal™

Catalog No. AC436040500
Change view
Click to view available options
Quantity:
50 mL
Packaging:
AcroSeal™ Glass Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC436040500 50 mL AcroSeal™ Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC436040500 Supplier Thermo Scientific Chemicals Supplier No. 436040500
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 112022-83-0 | C18H20BNO | 277.17 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 112022-83-0 , 108-88-3
Molecular Formula C18H20BNO
Molecular Weight (g/mol) 277.17
MDL Number MFCD00078440
InChI Key VMKAFJQFKBASMU-QGZVFWFLSA-N
Synonym r-2-methyl-cbs-oxazaborolidine, r-methyl oxazaborolidine, r-+-2-methyl-cbs-oxazaborolidine, r-5,5-diphenyl-2-methyl-3,4-propano-1,3,2-oxazaborolidine, r-1-methyl-3,3-diphenylhexahydropyrrolo 1,2-c 1,3,2 oxazaborole, r-me-cbs catalyst, r-3,3-diphenyl-1-methylpyrrolidino 1,2-c-1,3,2-oxazaborole, r-me cbs, r-tetrahydro-1-methyl-3,3-diphenyl-1h,3h-pyrrolo 1,2-c 1,3,2 oxazaborole, corey's catalyst
PubChem CID 9838490
IUPAC Name (3aR)-1-methyl-3,3-diphenyl-3a,4,5,6-tetrahydropyrrolo[1,2-c][1,3,2]oxazaborole
SMILES [H][C@]12CCCN1B(C)OC2(C1=CC=CC=C1)C1=CC=CC=C1

Specifications

CAS Min % 26.26, 67.91
CAS Max % 32.09, 73.74
Color Amber to Colorless
Density 0.9500g/mL
Boiling Point 111.0°C
Flash Point 4°C
Packaging AcroSeal™ Glass Bottle
Quantity 50 mL
Specific Gravity 0.95
Solubility Information Solubility in water: reacts
Formula Weight 277.17
Physical Form Liquid
Chemical Name or Material (R)-(+)-2-Methyl-CBS-oxazaborolidine
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Causes serious eye damage.
Harmful if swallowed.
May be fatal if swallowed and enters airways.
May cause drowsiness or dizziness.
May cause damage to organs through prolonged or repeated exposure.
Caus
Precautionary Statement GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
IF INHALED: Remov

RUO – Research Use Only