accessibility menu, dialog, popup

UserName

BNO(R)CH3H

CAS RN 112022-83-0

IUPAC Name: (3aR)-1-methyl-3,3-diphenyl-hexahydropyrrolo[1,2-c][1,3,2]oxazaborole
Synonyms: (3aR)-1-methyl-3,3-diphenyl-tetrahydropyrrolo[1,2-c][1,3,2]oxazaborole
Molecular Weight (g/mol): 277.17
Molecular Formula: C18H20BNO
InChi Key: VMKAFJQFKBASMU-QGZVFWFLSA-N
SMILES: [H][C@]12CCCN1B(C)OC2(C1=CC=CC=C1)C1=CC=CC=C1
Quantity 
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
Boiling Point 
  • (3)
Physical Form 
  • (2)
  • (1)
Percent Purity 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (2)
  • (4)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (3)
  • (2)
  • (1)
  • (2)