missing translation for 'onlineSavingsMsg'
Learn More

(R)-(+)-2-Methyl-2-propanesulfinamide, 98%

Catalog No. AAH2772414
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH2772414 25 g
AAH2772403 1 g
AAH2772406 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAH2772414 Supplier Thermo Scientific Chemicals Supplier No. H2772414
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 196929-78-9 | C4H11NOS | 121.20 g/mol

(R)-(+)-2-Methyl-2-propanesulfinamide is a chiral ligand, which is used in pharmaceutical compositions. Further, it is used in the preparation of beta-chloro sulfinamides in the synthesis of chiral azridines. It is involved in the preparation of organocatalyst for enantioselective reduction of imines. It serves as a reagent for synthesizing chiral amines. In addition to this, it is converted into P,N-sulfinyl imine ligands through condensation with aldehydes and ketones which undergoes iridium-catalyzed asymmetric hydrogenation of olefins.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
(R)-(+)-2-Methyl-2-propanesulfinamide is a chiral ligand, which is used in pharmaceutical compositions. Further, it is used in the preparation of beta-chloro sulfinamides in the synthesis of chiral azridines. It is involved in the preparation of organocatalyst for enantioselective reduction of imines. It serves as a reagent for synthesizing chiral amines. In addition to this, it is converted into P,N-sulfinyl imine ligands through condensation with aldehydes and ketones which undergoes iridium-catalyzed asymmetric hydrogenation of olefins.

Solubility
Soluble in chloroform, methanol, tetrahydrofuran, dichloromethane, dimethyl sulfoxide and most organic solvents.

Notes
Incompatible with strong oxidizing agents, strong acids and bases. Store in a cool place.

Chemical Identifiers

CAS 196929-78-9
Molecular Formula C4H11NOS
Molecular Weight (g/mol) 121.20
MDL Number MFCD05861479
InChI Key CESUXLKAADQNTB-UHFFFAOYNA-N
Synonym r-+-2-methyl-2-propanesulfinamide, r-2-methylpropane-2-sulfinamide, r-+-t-butylsulfinamide, r-+-tert-butanesulfinamide, r-+-tert-butylsulfinamide, r-2-methyl-2-propanesulfinamide, r-tert-butanesulfinamide, r-+-tert-butyl sulphinamide, r-tert-butylsulfinamide, r-+-tert-butyl sulfinamide
PubChem CID 10964479
IUPAC Name 2-methylpropane-2-sulfinamide
SMILES CC(C)(C)S(N)=O

Specifications

Melting Point ∼100°C to 105°C
Quantity 25 g
Solubility Information Soluble in chloroform,methanol,tetrahydrofuran,dichloromethane,dimethyl sulfoxide and most organic solvents.
Formula Weight 121.2
Percent Purity 98%
Chemical Name or Material (R)-(+)-2-Methyl-2-propanesulfinamide
Hazard Category H315-H319-H335
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Keep cold

RUO – Research Use Only