missing translation for 'onlineSavingsMsg'
Learn More

(R)-(+)-2-Amino-3-methyl-1,1-diphenyl-1-butanol 98.0+%, TCI America™

Catalog No. A25331G
Change view
Click to view available options
Quantity:
1 g
Catalog No. Quantity
A25331G 1 g
1 options

Catalog No. A25331G

Supplier: TCI America A25331G

Only null left

Chemical Identifiers

CAS 86695-06-9
Molecular Formula C17H21NO
Molecular Weight (g/mol) 255.361
MDL Number MFCD01318247
InChI Key LNQVZZGGOZBOQS-MRXNPFEDSA-N
PubChem CID 702733
IUPAC Name (2R)-2-amino-3-methyl-1,1-diphenylbutan-1-ol
SMILES CC(C)C(C(C1=CC=CC=C1)(C2=CC=CC=C2)O)N

Specifications

Melting Point 98°C
Color White-Yellow
Quantity 1 g
Formula Weight 255.36
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material (R)-(+)-2-Amino-3-methyl-1,1-diphenyl-1-butanol