missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(R)-(-)-2,3-Diaminopropionic Acid Hydrochloride 98.0+%, TCI America™
Specifications
Specifications
| Melting Point | 240°C |
| Color | White-Yellow |
| Quantity | 100 mg |
| Formula Weight | 140.57 |
| Percent Purity | ≥98.0% (N) |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | (R)-(-)-2,3-Diaminopropionic Acid Hydrochloride |
Safety and Handling
| TSCA | No |