missing translation for 'onlineSavingsMsg'
Learn More

(R)-(-)-2,2-Dimethyl-1,3-dioxolan-4-ylmethyl p-Toluenesulfonate 98.0+%, TCI America™

Catalog No. D25491G
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D25491G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D25491G Supplier TCI America Supplier No. D25491G
Only null left

Chemical Identifiers

CAS 23788-74-1
Molecular Formula C13H18O5S
Molecular Weight (g/mol) 286.342
MDL Number MFCD00063233
InChI Key SRKDUHUULIWXFT-LLVKDONJSA-N
Synonym r-2,2-dimethyl-1,3-dioxolan-4-yl methyl 4-methylbenzenesulfonate, r---2,2-dimethyl-1,3-dioxolan-4-ylmethyl p-toluenesulfonate, 4r-2,2-dimethyl-1,3-dioxolan-4-yl methyl 4-methylbenzenesulfonate, d---1,2-isopropylideneglycerol 3-p-toluenesulfonate, r-2,2-dimethyl-1,3-dioxolane-4-methanol p-toluenesulfonate, r---2,2-dimethyl-1,3-dioxolane-4-ylmethyl p-toluenesulfonate, r---2,2-dimethyl-4-hydroxymethyl-1,3-dioxolane-p-toluenesulfonate, d-alpha,beta-isopropylideneglycerol-gamma-tosylate, pubchem6799, 2,2-dimethyl-1,3-dioxolan-4-yl methyl 4-methylbenzenesulfonate #
PubChem CID 6993835
IUPAC Name [(4R)-2,2-dimethyl-1,3-dioxolan-4-yl]methyl 4-methylbenzenesulfonate
SMILES CC1=CC=C(C=C1)S(=O)(=O)OCC2COC(O2)(C)C

Specifications

Color White-Yellow
Quantity 1 g
Formula Weight 286.34
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Lumps
Chemical Name or Material (R)-(-)-2,2-Dimethyl-1,3-dioxolan-4-ylmethyl p-Toluenesulfonate
TSCA No
Recommended Storage Refrigerator