missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(R)-1-(tert-Butoxycarbonyl)-2-azetidinemethanol 98.0+%, TCI America™
Spécifications
Spécifications
| Color | Yellow |
| Quantity | 1 g |
| Formula Weight | 187.24 |
| Percent Purity | ≥98.0% (GC) |
| Physical Form | Liquid |
| Chemical Name or Material | (R)-1-(tert-Butoxycarbonyl)-2-azetidinemethanol |
Sécurité et manipulation
| TSCA | No |