missing translation for 'onlineSavingsMsg'
Learn More

(R)-1-Phenyl-1,2-ethanediyl Bis[4-(trans-4-pentylcyclohexyl)benzoate] 98.0+%, TCI America™

Supplier:  TCI America P21501G

 View more versions of this product

Catalog No. P21501G

Only null left
Explore available promotions

Description

Description

Specifications

Specifications

(R)-1-Phenyl-1,2-ethanediyl Bis[4-(trans-4-pentylcyclohexyl)benzoate]
132°C
C44H58O4
1 g
CCCCCC1CCC(CC1)C2=CC=C(C=C2)C(=O)OCC(C3=CC=CC=C3)OC(=O)C4=CC=C(C=C4)C5CCC(CC5)CCCCC
650.944
650.94
Crystalline Powder
154102-21-3
White
MFCD26961137
KTIVHFRVDVVCHK-BAFIUCHISA-N
[(2R)-2-[4-(4-pentylcyclohexyl)benzoyl]oxy-2-phenylethyl] 4-(4-pentylcyclohexyl)benzoate
57719999
≥98.0% (HPLC)

Chemical Identifiers

154102-21-3
650.944
KTIVHFRVDVVCHK-BAFIUCHISA-N
[(2R)-2-[4-(4-pentylcyclohexyl)benzoyl]oxy-2-phenylethyl] 4-(4-pentylcyclohexyl)benzoate
C44H58O4
MFCD26961137
57719999
CCCCCC1CCC(CC1)C2=CC=C(C=C2)C(=O)OCC(C3=CC=CC=C3)OC(=O)C4=CC=C(C=C4)C5CCC(CC5)CCCCC
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

TSCA : No

SDS
Documents

Documents

Promotions

Promotions