missing translation for 'onlineSavingsMsg'
Learn More

(R)-1-Phenyl-1,2-ethanediyl Bis[4-(trans-4-pentylcyclohexyl)benzoate] 98.0+%, TCI America™

Catalog No. p-7135430
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P2150200MG 200 mg
P21501G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P2150200MG Supplier TCI America Supplier No. P2150200MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 154102-21-3
Molecular Formula C44H58O4
Molecular Weight (g/mol) 650.944
MDL Number MFCD26961137
InChI Key KTIVHFRVDVVCHK-BAFIUCHISA-N
PubChem CID 57719999
IUPAC Name [(2R)-2-[4-(4-pentylcyclohexyl)benzoyl]oxy-2-phenylethyl] 4-(4-pentylcyclohexyl)benzoate
SMILES CCCCCC1CCC(CC1)C2=CC=C(C=C2)C(=O)OCC(C3=CC=CC=C3)OC(=O)C4=CC=C(C=C4)C5CCC(CC5)CCCCC

Specifications

Melting Point 132°C
Color White
Quantity 200 mg
Formula Weight 650.94
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material (R)-1-Phenyl-1,2-ethanediyl Bis[4-(trans-4-pentylcyclohexyl)benzoate]