accessibility menu, dialog, popup

UserName

CAS RN 154102-21-3

IUPAC Name: 2-[4-(4-pentylcyclohexyl)benzoyloxy]-2-phenylethyl 4-(4-pentylcyclohexyl)benzoate
Synonyms: 2-[4-(4-pentylcyclohexyl)benzoyloxy]-2-phenylethyl 4-(4-pentylcyclohexyl)benzoate
Molecular Weight (g/mol): 650.94
Molecular Formula: C44H58O4
InChi Key: KTIVHFRVDVVCHK-UHFFFAOYNA-N
SMILES: CCCCCC1CCC(CC1)C1=CC=C(C=C1)C(=O)OCC(OC(=O)C1=CC=C(C=C1)C1CCC(CCCCC)CC1)C1=CC=CC=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results