missing translation for 'onlineSavingsMsg'
Learn More

(R)-(-)-1-Methyl-3-pyrrolidinol, 99%

Catalog No. AC439440050
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC439440050 5 g
AC439440010 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC439440050 Supplier Thermo Scientific Chemicals Supplier No. 439440050
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 104641-60-3 | C5H11NO | 101.15 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 104641-60-3
Molecular Formula C5H11NO
Molecular Weight (g/mol) 101.15
MDL Number MFCD03788747
InChI Key FLVFPAIGVBQGET-UHFFFAOYNA-N
Synonym 3r-1-methylpyrrolidin-3-ol, r---1-methyl-3-pyrrolidinol, r-3-hydroxy-1-methyl-pyrrolidine, r-1-methyl-3-pyrrolidinol, r-1-methylpyrrolidin-3-ol, r-3-hydroxy-1-methylpyrrolidine, r---1-methyl-3-hydroxypyrrolidine, 3-pyrrolidinol, 1-methyl-, 3r, r-1-methyl-pyrrolidin-3-ol, unii-6hvw4ew50x
PubChem CID 6951332
IUPAC Name (3R)-1-methylpyrrolidin-3-ol
SMILES CN1CCC(O)C1

Specifications

CAS Min % 98.5
CAS Max % 100.0
Color Brown to Colorless
Density 0.9210g/mL
Boiling Point 50°C to 52°C (1.0 mmHg)
Flash Point 70°C
Infrared Spectrum Authentic
Assay Percent Range 99%
Refractive Index 1.464
Quantity 5 g
Specific Gravity 0.921
Specific Rotation -7
Formula Weight 101.15
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material (R)-(-)-1-Methyl-3-pyrrolidinol
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Recommended Storage May darken on storage

RUO – Research Use Only