missing translation for 'onlineSavingsMsg'
Learn More

(R)-1-Acetyl-3-pyrrolidinol 98.0+%, TCI America™

Catalog No. A20491G
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A20491G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A20491G Supplier TCI America Supplier No. A20491G
Only null left

Chemical Identifiers

CAS 916733-17-0
Molecular Formula C6H11NO2
Molecular Weight (g/mol) 129.159
InChI Key ODHQVFPGHQBQSY-ZCFIWIBFSA-N
Synonym (R)-1-Acetyl-3-hydroxypyrrolidine
PubChem CID 10486876
IUPAC Name 1-[(3R)-3-hydroxypyrrolidin-1-yl]ethanone
SMILES CC(=O)N1CCC(C1)O

Specifications

Melting Point 83°C
Color White-Yellow
Quantity 1 g
Formula Weight 129.16
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material (R)-1-Acetyl-3-pyrrolidinol